Product Details

SNP ID
rs17045452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:53984047 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGACACAAGGAGGCTTGTGGTTTA[A/G]AAAGATGACTCTGGGCATTTTCATC
Phenotype
MIM: 102595
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACYP2 PubMed Links

Gene Details

Gene
ACYP2
Gene Name
acylphosphatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320586.1 Intron NP_001307515.1
NM_001320587.1 Intron NP_001307516.1
NM_001320588.1 Intron NP_001307517.1
NM_001320589.1 Intron NP_001307518.1
NM_001320590.1 Intron NP_001307519.1
NM_138448.3 Intron NP_612457.1
XM_017005406.1 Intron XP_016860895.1
XM_017005407.1 Intron XP_016860896.1
XM_017005408.1 Intron XP_016860897.1
XM_017005409.1 Intron XP_016860898.1
XM_017005410.1 Intron XP_016860899.1
XM_017005411.1 Intron XP_016860900.1
XM_017005412.1 Intron XP_016860901.1
XM_017005413.1 Intron XP_016860902.1

View Full Product Details