Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001024383.1 | 218 | Intron | NP_001019554.1 | ||
NM_014903.5 | 218 | Intron | NP_055718.4 | ||
XM_005269215.3 | 218 | Intron | XP_005269272.1 | ||
XM_011538940.2 | 218 | Intron | XP_011537242.1 | ||
XM_011538941.2 | 218 | Intron | XP_011537243.1 | ||
XM_011538944.2 | 218 | Intron | XP_011537246.1 | ||
XM_017020164.1 | 218 | Intron | XP_016875653.1 | ||
XM_017020165.1 | 218 | Intron | XP_016875654.1 | ||
XM_017020166.1 | 218 | UTR 5 | XP_016875655.1 | ||
XM_017020167.1 | 218 | UTR 5 | XP_016875656.1 | ||
XM_017020168.1 | 218 | Intron | XP_016875657.1 | ||
XM_017020169.1 | 218 | Intron | XP_016875658.1 | ||
XM_017020170.1 | 218 | Intron | XP_016875659.1 | ||
XM_017020171.1 | 218 | Intron | XP_016875660.1 | ||
XM_017020172.1 | 218 | Intron | XP_016875661.1 | ||
XM_017020173.1 | 218 | Intron | XP_016875662.1 | ||
XM_017020174.1 | 218 | Intron | XP_016875663.1 | ||
XM_017020175.1 | 218 | Intron | XP_016875664.1 |