Product Details

SNP ID
rs17500109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:101393655 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTATTCTTTGTGATAAACCAGAGG[C/G]ATCAGGCTTAACCAAGTGCTGAACT
Phenotype
MIM: 603425 MIM: 612822
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARL1 PubMed Links

Gene Details

Gene
ARL1
Gene Name
ADP ribosylation factor like GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177.5 2706 UTR 3 NP_001168.1
NM_001301068.1 2706 UTR 3 NP_001287997.1
Gene
UTP20
Gene Name
UTP20, small subunit processome component
There are no transcripts associated with this gene.

View Full Product Details