Product Details

SNP ID
rs36013132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109573424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCTGGCGGCGCCGAAGCACCC[GC/TT]GCAGGCCAAGACGGCTCCCCAGGC
Phenotype
MIM: 607568 MIM: 251170
Polymorphism
GC/TT, Multiple nucleotide polymorphism
Allele Nomenclature
Literature Links
MMAB PubMed Links

Gene Details

Gene
MMAB
Gene Name
methylmalonic aciduria (cobalamin deficiency) cblB type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052845.3 140 Silent Mutation NP_443077.1
XM_011538267.2 140 Intron XP_011536569.1
XM_011538268.2 140 Intron XP_011536570.1
XM_011538269.2 140 Intron XP_011536571.1
Gene
MVK
Gene Name
mevalonate kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000431.3 140 Intron NP_000422.1
NM_001114185.2 140 Intron NP_001107657.1
NM_001301182.1 140 Intron NP_001288111.1
XM_011538372.2 140 Silent Mutation GCC,GCG A47A XP_011536674.2
XM_017019313.1 140 Intron XP_016874802.1
XM_017019314.1 140 Intron XP_016874803.1

View Full Product Details