Product Details

SNP ID
rs17854373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:97644738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTGGGCCGCGGCTTGCGCCGGC[G/T]CCCGCAAAAAGCGGAGGAGCCCGCC
Phenotype
MIM: 610323
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MTDH PubMed Links

Gene Details

Gene
MTDH
Gene Name
metadherin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178812.3 767 Missense Mutation GCC,TCC A78S NP_848927.2
XM_005251099.3 767 Missense Mutation GCC,TCC A78S XP_005251156.1
XM_011517367.2 767 Missense Mutation GCC,TCC A78S XP_011515669.1
XM_011517368.2 767 Missense Mutation GCC,TCC A78S XP_011515670.1
XM_011517369.2 767 Missense Mutation GCC,TCC A78S XP_011515671.1
XM_011517370.2 767 Missense Mutation GCC,TCC A78S XP_011515672.1
XM_017013966.1 767 Missense Mutation GCC,TCC A78S XP_016869455.1
XM_017013967.1 767 Missense Mutation GCC,TCC A78S XP_016869456.1
XM_017013968.1 767 Missense Mutation GCC,TCC A78S XP_016869457.1

View Full Product Details