Product Details

SNP ID
rs16838424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:206658901 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAAAAGGATGGAAAAAGAAGCTA[C/G]AAAAATAACTTAGTAGACATATGCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DYTN PubMed Links

Gene Details

Gene
DYTN
Gene Name
dystrotelin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001093730.1 Intron NP_001087199.1
Gene
LOC200726
Gene Name
hCG1657980
There are no transcripts associated with this gene.

View Full Product Details