Product Details
- SNP ID
-
rs17730834
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:206660137 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGATTGTGAGTGAAAGCATTAAGT[A/G]AATATTTTGAGACTTTTAAAAATAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DYTN
PubMed Links
Gene Details
- Gene
- DYTN
- Gene Name
- dystrotelin
View Full Product Details