Product Details

SNP ID
rs13447751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:94418692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATATTAGATTTCAAGTTCAATAA[A/G]GATGTGGGCAGATCTGCCTAAGAAT
Phenotype
MIM: 600814
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRE11A PubMed Links

Gene Details

Gene
MRE11A
Gene Name
MRE11 homolog A, double strand break repair nuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005590.3 3464 UTR 3 NP_005581.2
NM_005591.3 3464 UTR 3 NP_005582.1
XM_005274008.3 3464 UTR 3 XP_005274065.1
XM_006718842.3 3464 UTR 3 XP_006718905.1
XM_011542837.2 3464 UTR 3 XP_011541139.1
XM_017017772.1 3464 UTR 3 XP_016873261.1
XM_017017773.1 3464 UTR 3 XP_016873262.1

View Full Product Details