Product Details

SNP ID
hCV33342413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130510178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACTGGTGGGGACTATGAAGAAT[A/G]GGTCTGAATTCAGAGAGCAGATGGA
Phenotype
MIM: 604355 MIM: 601029
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPG2 PubMed Links

Gene Details

Gene
COPG2
Gene Name
coatomer protein complex subunit gamma 2
There are no transcripts associated with this gene.

Gene
MEST
Gene Name
mesoderm specific transcript
There are no transcripts associated with this gene.

View Full Product Details