Product Details

SNP ID
rs17004580
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:43576676 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATCTTCTGTATTCCTACAAATAC[G/T]CTGCAAAATCTTGAAAAATTAAATT
Phenotype
MIM: 604554
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
HSF2BP PubMed Links

Gene Details

Gene
HSF2BP
Gene Name
heat shock transcription factor 2 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007031.1 Intron NP_008962.1
XM_011529443.2 Intron XP_011527745.2
XM_011529445.2 Intron XP_011527747.2
XM_011529446.2 Intron XP_011527748.1
XM_011529447.2 Intron XP_011527749.1
XM_017028267.1 Intron XP_016883756.1
XM_017028268.1 Intron XP_016883757.1
XM_017028269.1 Intron XP_016883758.1
XM_017028270.1 Intron XP_016883759.1
XM_017028271.1 Intron XP_016883760.1
XM_017028272.1 Intron XP_016883761.1

View Full Product Details