Product Details

SNP ID
rs16953638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:96884739 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATAAAGTTGAGATCCTCTGGTATC[A/C]ACTCATCACAGCTGTGCACGATGAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HSP90AB6P PubMed Links

Gene Details

Gene
HSP90AB6P
Gene Name
heat shock protein 90 alpha family class B member 6, pseudogene
There are no transcripts associated with this gene.

View Full Product Details