Product Details

SNP ID
rs7769422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:24673685 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAATTCTCTTATAATTTATTAAGA[G/T]TAAGTTAACCCCTTAAGAAAAGTTT
Phenotype
MIM: 615652 MIM: 605764
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACOT13 PubMed Links

Gene Details

Gene
ACOT13
Gene Name
acyl-CoA thioesterase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160094.1 Intron NP_001153566.1
NM_018473.3 Intron NP_060943.1
Gene
TDP2
Gene Name
tyrosyl-DNA phosphodiesterase 2
There are no transcripts associated with this gene.

View Full Product Details