Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_020854.3 | Intron | NP_065905.2 | |||
XM_005266730.2 | Intron | XP_005266787.1 | |||
XM_011526110.1 | Intron | XP_011524412.1 | |||
XM_011526111.1 | Intron | XP_011524413.1 | |||
XM_011526112.1 | Intron | XP_011524414.1 | |||
XM_011526114.1 | Intron | XP_011524416.1 | |||
XM_011526117.2 | Intron | XP_011524419.1 | |||
XM_017025882.1 | Intron | XP_016881371.1 | |||
XM_017025883.1 | Intron | XP_016881372.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_012327.5 | Intron | NP_036459.1 | |||
NM_176787.4 | Intron | NP_789744.1 | |||
XM_011525889.1 | Intron | XP_011524191.1 | |||
XM_011525890.1 | Intron | XP_011524192.1 | |||
XM_011525891.1 | Intron | XP_011524193.1 | |||
XM_011525892.1 | Intron | XP_011524194.1 | |||
XM_011525893.1 | Intron | XP_011524195.1 | |||
XM_011525894.1 | Intron | XP_011524196.1 | |||
XM_011525895.1 | Intron | XP_011524197.1 | |||
XM_011525896.1 | Intron | XP_011524198.1 | |||
XM_011525898.1 | Intron | XP_011524200.1 | |||
XM_017025685.1 | Intron | XP_016881174.1 | |||
XM_017025686.1 | Intron | XP_016881175.1 |