Product Details

SNP ID
rs17853053
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:161098547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCCCGGGGCCCATGGCGGTGGC[C/T]GTGCCCCCGGGTCGGGCCGCAGGCT
Phenotype
MIM: 609607 MIM: 613466
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHDC9 PubMed Links

Gene Details

Gene
KLHDC9
Gene Name
kelch domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007255.2 157 Silent Mutation GCC,GCT A4A NP_001007256.1
NM_152366.4 157 Silent Mutation GCC,GCT A4A NP_689579.3
Gene
NECTIN4
Gene Name
nectin cell adhesion molecule 4
There are no transcripts associated with this gene.

Gene
PFDN2
Gene Name
prefoldin subunit 2
There are no transcripts associated with this gene.

View Full Product Details