Product Details

SNP ID
rs17181988
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:161038634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCGAAGAGCCGGGCCCGTCCGGAG[A/G]CTAGGAGTGAACGGAGTTCAGGAAG
Phenotype
MIM: 614264 MIM: 616041 MIM: 191523
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP30 PubMed Links

Gene Details

Gene
ARHGAP30
Gene Name
Rho GTPase activating protein 30
There are no transcripts associated with this gene.

Gene
TSTD1
Gene Name
thiosulfate sulfurtransferase like domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113205.1 145 Missense Mutation GCC,GTC A17V NP_001106676.1
NM_001113206.1 145 Intron NP_001106677.1
NM_001113207.1 145 Missense Mutation GCC,GTC A17V NP_001106678.1
Gene
USF1
Gene Name
upstream transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276373.1 145 Intron NP_001263302.1
NM_007122.4 145 Intron NP_009053.1
NM_207005.2 145 Intron NP_996888.1

View Full Product Details