Product Details

SNP ID
rs16972973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42274960 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGTTGCAATCAGAACTCTTAACA[C/T]GTGAATTTCAGGCATGTTGCTTTCC
Phenotype
MIM: 104180
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GANC PubMed Links

Gene Details

Gene
GANC
Gene Name
glucosidase alpha, neutral C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301409.1 Intron NP_001288338.1
NM_001301410.1 Intron NP_001288339.1
NM_198141.2 Intron NP_937784.2
Gene
TMEM87A
Gene Name
transmembrane protein 87A
There are no transcripts associated with this gene.

View Full Product Details