Product Details

SNP ID
rs17147157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:37378060 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATGTGAAAAACAATTTTAGACC[A/G]GAATAGCAAGGTGAGTAAGGCCAAA
Phenotype
MIM: 300935
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRRG1 PubMed Links

Gene Details

Gene
PRRG1
Gene Name
proline rich and Gla domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000950.2 Intron NP_000941.1
NM_001142395.1 Intron NP_001135867.1
NM_001173486.1 Intron NP_001166957.1
NM_001173489.1 Intron NP_001166960.1
NM_001173490.1 Intron NP_001166961.1

View Full Product Details