Product Details

SNP ID
rs34291123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:158731813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTATTTAAAACTGAAAATTAAC[A/G]TTTTGACAACTCAAGAGTGTCTGAC
Phenotype
MIM: 616691
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ESYT2 PubMed Links

Gene Details

Gene
ESYT2
Gene Name
extended synaptotagmin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020728.2 5142 UTR 3 NP_065779.1

View Full Product Details