Product Details

SNP ID
rs17307294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:14526924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCCTGGAATGTAGACTAATGGT[A/G]AGCATACAGATTTGTTTTTAATTTG
Phenotype
MIM: 400028
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NLGN4Y PubMed Links

Gene Details

Gene
NLGN4Y
Gene Name
neuroligin 4, Y-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164238.1 Intron NP_001157710.1
NM_001206850.1 Intron NP_001193779.1
NM_014893.4 Intron NP_055708.3
XM_006724874.2 Intron XP_006724937.1
XM_011531424.2 Intron XP_011529726.1
XM_011531425.2 Intron XP_011529727.1
XM_011531426.2 Intron XP_011529728.1
XM_011531427.1 Intron XP_011529729.1
XM_011531428.1 Intron XP_011529730.1
XM_011531429.2 Intron XP_011529731.1
XM_011531430.2 Intron XP_011529732.1
XM_017030034.1 Intron XP_016885523.1
XM_017030035.1 Intron XP_016885524.1
XM_017030036.1 Intron XP_016885525.1
XM_017030037.1 Intron XP_016885526.1
XM_017030038.1 Intron XP_016885527.1
XM_017030039.1 Intron XP_016885528.1
XM_017030040.1 Intron XP_016885529.1
XM_017030041.1 Intron XP_016885530.1

View Full Product Details