Product Details

SNP ID
rs17767418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30479276 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTACTTCTGCCCTTTTATTTTAAC[C/T]TTCACTTCACCATGAATGAGAATAC
Phenotype
MIM: 603102 MIM: 604026
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPD PubMed Links

Gene Details

Gene
CPD
Gene Name
carboxypeptidase D
There are no transcripts associated with this gene.

Gene
GOSR1
Gene Name
golgi SNAP receptor complex member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007024.1 Intron NP_001007025.1
NM_001007025.1 Intron NP_001007026.1
NM_004871.2 Intron NP_004862.1
XM_005258070.4 Intron XP_005258127.1
XM_005258071.4 Intron XP_005258128.1
XM_005258072.2 Intron XP_005258129.1
XM_017025374.1 Intron XP_016880863.1
XM_017025375.1 Intron XP_016880864.1
XM_017025376.1 Intron XP_016880865.1
XM_017025377.1 Intron XP_016880866.1

View Full Product Details