Product Details
- SNP ID
-
rs17272603
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:47823315 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGCCACAGAGAGGAGGTGTGATGC[A/G]GTTTTCCCAAAAGGAACGAGCGAGG
- Phenotype
-
MIM: 602225
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CRX
PubMed Links
Gene Details
- Gene
- CRX
- Gene Name
- cone-rod homeobox
There are no transcripts associated with this gene.
View Full Product Details