Product Details

SNP ID
rs17586936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:44555196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGCTGTGGTTAGTGCGTGAGAT[A/G]AAAGTGCAGGATCCTATAAATAGAT
Phenotype
MIM: 603910 MIM: 610844
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF3J PubMed Links

Gene Details

Gene
EIF3J
Gene Name
eukaryotic translation initiation factor 3 subunit J
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284335.1 Intron NP_001271264.1
NM_001284336.1 Intron NP_001271265.1
NM_003758.3 Intron NP_003749.2
Gene
SPG11
Gene Name
spastic paraplegia 11 (autosomal recessive)
There are no transcripts associated with this gene.

View Full Product Details