Product Details

SNP ID
rs17515108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:44563817 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGATACTGTATTTTGAGTGAGGG[A/C]TGTCAGCTCTGAAGTACCACTTACT
Phenotype
MIM: 603910 MIM: 610844
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EIF3J PubMed Links

Gene Details

Gene
EIF3J
Gene Name
eukaryotic translation initiation factor 3 subunit J
There are no transcripts associated with this gene.

Gene
SPG11
Gene Name
spastic paraplegia 11 (autosomal recessive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160227.1 Intron NP_001153699.1
NM_025137.3 Intron NP_079413.3
XM_006720700.1 Intron XP_006720763.1
XM_006720701.3 Intron XP_006720764.1
XM_017022634.1 Intron XP_016878123.1
XM_017022635.1 Intron XP_016878124.1
XM_017022636.1 Intron XP_016878125.1

View Full Product Details