Product Details

SNP ID
rs113994043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184135551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGCCGATAGCTTCGATCGCCGC[G/T]TCTTCCCCATCTCCAAGGACCAGCC
Phenotype
MIM: 603945
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EIF2B5 PubMed Links

Gene Details

Gene
EIF2B5
Gene Name
eukaryotic translation initiation factor 2B subunit epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003907.2 530 Missense Mutation GTC,TTC V56F NP_003898.2
XM_011513265.1 530 Intron XP_011511567.1
XM_011513266.2 530 Intron XP_011511568.1

View Full Product Details