Product Details

SNP ID
rs16958576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:32768949 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTTCACATGAGCTCTTGACTGAA[A/G]GACACGATCCTATCAGAGAAAGGTA
Phenotype
MIM: 136535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FMN1 PubMed Links

Gene Details

Gene
FMN1
Gene Name
formin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103184.3 8529 UTR 3 NP_001096654.1
NM_001277313.1 8529 UTR 3 NP_001264242.1
NM_001277314.1 8529 Intron NP_001264243.1
XM_011521504.2 8529 UTR 3 XP_011519806.1
XM_011521505.2 8529 Intron XP_011519807.1
XM_011521506.2 8529 UTR 3 XP_011519808.1
XM_011521507.2 8529 Intron XP_011519809.1
XM_011521509.2 8529 UTR 3 XP_011519811.1
XM_011521511.2 8529 UTR 3 XP_011519813.1
XM_017022130.1 8529 UTR 3 XP_016877619.1
XM_017022131.1 8529 Intron XP_016877620.1
XM_017022132.1 8529 UTR 3 XP_016877621.1
XM_017022133.1 8529 UTR 3 XP_016877622.1
XM_017022134.1 8529 UTR 3 XP_016877623.1
XM_017022135.1 8529 UTR 3 XP_016877624.1

View Full Product Details