Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001166301.1 | 180 | Silent Mutation | CTC,CTG | L29L | NP_001159773.1 |
NM_024612.4 | 180 | Silent Mutation | CTC,CTG | L29L | NP_078888.4 |
XM_006722083.2 | 180 | Silent Mutation | CTC,CTG | L29L | XP_006722146.1 |
XM_011525253.2 | 180 | Silent Mutation | CTC,CTG | L29L | XP_011523555.1 |
XM_011525254.2 | 180 | UTR 5 | XP_011523556.1 | ||
XM_017025080.1 | 180 | Silent Mutation | CTC,CTG | L29L | XP_016880569.1 |
XM_017025081.1 | 180 | UTR 5 | XP_016880570.1 | ||
XM_017025082.1 | 180 | UTR 5 | XP_016880571.1 | ||
XM_017025083.1 | 180 | UTR 5 | XP_016880572.1 |