Product Details

SNP ID
rs17344841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:19410267 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTAACAAAAACTGCAAACCAGCA[C/T]GTGCTCCCACTCTTATATGTGTACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AEBP2 PubMed Links

Gene Details

Gene
AEBP2
Gene Name
AE binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114176.1 Intron NP_001107648.1
NM_001267043.1 Intron NP_001253972.1
NM_153207.4 Intron NP_694939.2
XM_006719031.3 Intron XP_006719094.1
XM_011520560.2 Intron XP_011518862.1
XM_017018806.1 Intron XP_016874295.1

View Full Product Details