Product Details

SNP ID
rs17763689
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53512648 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAGTGACCAGTCTGCAAGGGAAT[A/G]TGGGACAACTCCTGTCCAGACCAAT
Phenotype
MIM: 606371 MIM: 604643 MIM: 605053
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ATF7 PubMed Links
Additional Information
For this assay, SNP(s) [rs80046306] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ATF7
Gene Name
activating transcription factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130060.1 6099 UTR 3 NP_001123532.1
NM_001206682.1 6099 Intron NP_001193611.1
NM_001206683.1 6099 Intron NP_001193612.1
NM_006856.2 6099 UTR 3 NP_006847.1
XM_005268587.3 6099 UTR 3 XP_005268644.1
XM_017018722.1 6099 UTR 3 XP_016874211.1
Gene
LOC100652999
Gene Name
uncharacterized LOC100652999
There are no transcripts associated with this gene.

Gene
NPFF
Gene Name
neuropeptide FF-amide peptide precursor
There are no transcripts associated with this gene.

Gene
TARBP2
Gene Name
TARBP2, RISC loading complex RNA binding subunit
There are no transcripts associated with this gene.

View Full Product Details