Product Details

SNP ID
rs16902858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:36251789 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTTTTGAATTTAAATTAGTTAAG[A/G]CCATCCCTCTTTCACTATTTTACAC
Phenotype
MIM: 615787 MIM: 616391
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NADK2 PubMed Links

Gene Details

Gene
NADK2
Gene Name
NAD kinase 2, mitochondrial
There are no transcripts associated with this gene.

Gene
RANBP3L
Gene Name
RAN binding protein 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161429.2 Intron NP_001154901.1
NM_001323273.1 Intron NP_001310202.1
NM_001323274.1 Intron NP_001310203.1
NM_001323275.1 Intron NP_001310204.1
NM_001323276.1 Intron NP_001310205.1
NM_001323277.1 Intron NP_001310206.1
NM_001323278.1 Intron NP_001310207.1
NM_001323279.1 Intron NP_001310208.1
NM_001323280.1 Intron NP_001310209.1
NM_145000.4 Intron NP_659437.3
XM_006714452.3 Intron XP_006714515.1
XM_006714453.3 Intron XP_006714516.1
XM_006714455.3 Intron XP_006714518.1

View Full Product Details