Product Details

SNP ID
rs17852680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40218055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGCTGCTGGTGGGCCCGGGGGC[G/T]GGGGCGGGCAGGCCGCTAGGGCGGG
Phenotype
MIM: 600137
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CNTD2 PubMed Links

Gene Details

Gene
CNTD2
Gene Name
cyclin N-terminal domain containing 2
There are no transcripts associated with this gene.

Gene
MAP3K10
Gene Name
mitogen-activated protein kinase kinase kinase 10
There are no transcripts associated with this gene.

Gene
TTC9B
Gene Name
tetratricopeptide repeat domain 9B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152479.5 345 Silent Mutation CCA,CCC P109P NP_689692.2
XM_017026368.1 345 Silent Mutation CCA,CCC P109P XP_016881857.1

View Full Product Details