Product Details

SNP ID
rs16978678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24556248 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGAAGTAAGTTCATTAGCTTTCC[C/T]CTCAACGTGCATGCTTCTCACATTC
Phenotype
MIM: 601062
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUCD1 PubMed Links

Gene Details

Gene
GUCD1
Gene Name
guanylyl cyclase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284251.1 Intron NP_001271180.1
NM_001284252.1 Intron NP_001271181.1
NM_001284253.1 Intron NP_001271182.1
NM_001284254.1 Intron NP_001271183.1
NM_001284255.1 Intron NP_001271184.1
NM_001284256.1 Intron NP_001271185.1
NM_001284257.1 Intron NP_001271186.1
NM_031444.3 Intron NP_113632.2
XM_005261761.1 Intron XP_005261818.1
Gene
SNRPD3
Gene Name
small nuclear ribonucleoprotein D3 polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278656.1 Intron NP_001265585.1
NM_004175.4 Intron NP_004166.1

View Full Product Details