Product Details

SNP ID
rs17764221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:173993030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTCTCCCCTATAGTCTGTACTC[C/T]GATTTGCCTAGTTATTTATTTATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C5orf47 PubMed Links

Gene Details

Gene
C5orf47
Gene Name
chromosome 5 open reading frame 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144954.1 Intron NP_001138426.1
XM_011534431.1 Intron XP_011532733.1
XM_017009028.1 Intron XP_016864517.1

View Full Product Details