Product Details
- SNP ID
-
rs17736143
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:61337449 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGCACCAGATTGTTATGCTTCTCCA[A/G]TACTTTTTTCTCGGACATATAAGAC
- Phenotype
-
MIM: 605807
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDH20
PubMed Links
Gene Details
- Gene
- CDH20
- Gene Name
- cadherin 20
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031891.3 |
|
Intron |
|
|
NP_114097.2 |
View Full Product Details