Product Details
- SNP ID
-
rs17032644
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:10304379 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCTCCCGGATTCCTTTTCTCATC[C/T]GGGTTTCGGAGGTGGACTCTGTCTG
- Phenotype
-
MIM: 600152
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SEC13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112628776] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SEC13
- Gene Name
- SEC13 homolog, nuclear pore and COPII coat complex component
View Full Product Details