Product Details

SNP ID
rs17306163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:53225139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGGGACAGTTCCACACTTGGGA[A/T]TGAGGACCTCTCATGCCATCCTGAG
Phenotype
MIM: 606781
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TKT PubMed Links

Gene Details

Gene
TKT
Gene Name
transketolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001064.3 2482 Intron NP_001055.1
NM_001135055.2 2482 UTR 3 NP_001128527.1
NM_001258028.1 2482 Intron NP_001244957.1
XM_011534054.1 2482 UTR 3 XP_011532356.1
XM_011534055.1 2482 Intron XP_011532357.1

View Full Product Details