Product Details

SNP ID
rs34462603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:141498313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTACCAAAGCCCACATCCCTTTCT[C/T]TACAAAGCTGCCTGTGAGATGATAT
Phenotype
MIM: 601589
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RASA2 PubMed Links

Gene Details

Gene
RASA2
Gene Name
RAS p21 protein activator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303245.1 Intron NP_001290174.1
NM_001303246.1 Intron NP_001290175.1
NM_006506.3 Intron NP_006497.2
XM_011513059.2 Intron XP_011511361.1
XM_017006968.1 Intron XP_016862457.1
XM_017006969.1 Intron XP_016862458.1
XM_017006970.1 Intron XP_016862459.1
XM_017006971.1 Intron XP_016862460.1
XM_017006972.1 Intron XP_016862461.1

View Full Product Details