Product Details

SNP ID
rs17149779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:126444664 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGTAAATTAGAGTAGACAATGCT[A/T]ATCTAGAAATCTTGATAGAAAAGAA
Phenotype
MIM: 601116
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GRM8 PubMed Links

Gene Details

Gene
GRM8
Gene Name
glutamate metabotropic receptor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000845.2 Intron NP_000836.2
NM_001127323.1 Intron NP_001120795.1
XM_006715938.3 Intron XP_006716001.1
XM_011516091.1 Intron XP_011514393.1
XM_011516092.2 Intron XP_011514394.1
XM_011516094.1 Intron XP_011514396.1
XM_011516095.2 Intron XP_011514397.1
XM_011516101.2 Intron XP_011514403.1
XM_011516102.2 Intron XP_011514404.1
XM_011516103.2 Intron XP_011514405.1
XM_011516108.2 Intron XP_011514410.1
XM_017012074.1 Intron XP_016867563.1
XM_017012075.1 Intron XP_016867564.1
XM_017012076.1 Intron XP_016867565.1
XM_017012077.1 Intron XP_016867566.1
XM_017012078.1 Intron XP_016867567.1
XM_017012079.1 Intron XP_016867568.1
XM_017012080.1 Intron XP_016867569.1

View Full Product Details