Product Details

SNP ID
rs17146980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5950581 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGTGGATGGGTGCCAAAGGACCT[A/G]TGTTGATTCCTTCCTTCCCTTTCAT
Phenotype
MIM: 601070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL15RA PubMed Links

Gene Details

Gene
IL15RA
Gene Name
interleukin 15 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243539.1 Intron NP_001230468.1
NM_001256765.1 Intron NP_001243694.1
NM_002189.3 Intron NP_002180.1
NM_172200.2 Intron NP_751950.2
XM_011519461.2 Intron XP_011517763.1
XM_011519462.2 Intron XP_011517764.1
XM_011519463.2 Intron XP_011517765.1
XM_011519464.2 Intron XP_011517766.1
XM_011519465.1 Intron XP_011517767.1
XM_011519466.2 Intron XP_011517768.1
XM_011519467.2 Intron XP_011517769.1
XM_011519468.1 Intron XP_011517770.1
XM_011519469.1 Intron XP_011517771.1
XM_011519470.2 Intron XP_011517772.1
XM_011519471.2 Intron XP_011517773.1
XM_011519472.2 Intron XP_011517774.1
XM_011519474.2 Intron XP_011517776.1
XM_011519475.2 Intron XP_011517777.1
XM_011519476.2 Intron XP_011517778.1
XM_011519477.2 Intron XP_011517779.1
XM_017016195.1 Intron XP_016871684.1
XM_017016196.1 Intron XP_016871685.1
XM_017016197.1 Intron XP_016871686.1
XM_017016198.1 Intron XP_016871687.1
XM_017016199.1 Intron XP_016871688.1
XM_017016200.1 Intron XP_016871689.1
XM_017016201.1 Intron XP_016871690.1
XM_017016202.1 Intron XP_016871691.1
XM_017016203.1 Intron XP_016871692.1

View Full Product Details