Product Details

SNP ID
rs17697849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:48323680 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTTCAAAATTACAAAGTTTTCC[A/G]TACATCTTAATCATTATAATTGAGG
Phenotype
MIM: 601158
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAPK8 PubMed Links

Gene Details

Gene
MAPK8
Gene Name
mitogen-activated protein kinase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278547.1 Intron NP_001265476.1
NM_001278548.1 Intron NP_001265477.1
NM_001323302.1 Intron NP_001310231.1
NM_001323320.1 Intron NP_001310249.1
NM_001323321.1 Intron NP_001310250.1
NM_001323322.1 Intron NP_001310251.1
NM_001323323.1 Intron NP_001310252.1
NM_001323324.1 Intron NP_001310253.1
NM_001323325.1 Intron NP_001310254.1
NM_001323326.1 Intron NP_001310255.1
NM_001323327.1 Intron NP_001310256.1
NM_001323328.1 Intron NP_001310257.1
NM_001323329.1 Intron NP_001310258.1
NM_001323330.1 Intron NP_001310259.1
NM_001323331.1 Intron NP_001310260.1
NM_139046.3 Intron NP_620634.1
NM_139049.3 Intron NP_620637.1

View Full Product Details