Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001198862.1 | Intron | NP_001185791.1 | |||
NM_018252.2 | Intron | NP_060722.2 | |||
XM_005273177.3 | Intron | XP_005273234.1 | |||
XM_011509717.2 | Intron | XP_011508019.1 | |||
XM_011509718.2 | Intron | XP_011508020.1 | |||
XM_017001634.1 | Intron | XP_016857123.1 | |||
XM_017001635.1 | Intron | XP_016857124.1 | |||
XM_017001636.1 | Intron | XP_016857125.1 | |||
XM_017001637.1 | Intron | XP_016857126.1 |