Product Details

SNP ID
rs17770612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:39571778 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTCAGCAAACAGTTTCTACTCTGT[A/G]ACATAAGCCCAGGAAAGTGAAGTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105375240 PubMed Links

Gene Details

Gene
LOC105375240
Gene Name
uncharacterized LOC105375240
There are no transcripts associated with this gene.

Gene
YAE1D1
Gene Name
Yae1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282446.1 Intron NP_001269375.1
NM_020192.4 Intron NP_064577.1

View Full Product Details