Product Details

SNP ID
rs16969914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78509414 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGGATGGCCAGAGCTAACAACCT[G/T]GCTTTACATGGCTATGGAAAACAGC
Phenotype
MIM: 614681 MIM: 147582
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HYKK PubMed Links

Gene Details

Gene
HYKK
Gene Name
hydroxylysine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013619.3 Intron NP_001013641.2
NM_001083612.1 Intron NP_001077081.1
XM_011521231.2 Intron XP_011519533.1
XM_017021922.1 Intron XP_016877411.1
Gene
IREB2
Gene Name
iron responsive element binding protein 2
There are no transcripts associated with this gene.

View Full Product Details