Product Details

SNP ID
rs17035053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:68161570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCATACAGAATATATTAGGGAAAG[G/T]TGGTGGTGAAGGAAATTCTCCAATA
Phenotype
MIM: 610729
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PNO1 PubMed Links

Gene Details

Gene
PNO1
Gene Name
partner of NOB1 homolog
There are no transcripts associated with this gene.

Gene
WDR92
Gene Name
WD repeat domain 92
There are no transcripts associated with this gene.

View Full Product Details