Product Details

SNP ID
rs16870921
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32824863 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTGTGGTTGCTCTGCCTTATTTT[G/T]AAATTTAATAGTCATTTTTATTACT
Phenotype
MIM: 600629 MIM: 170261
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
HLA-DOB PubMed Links
Additional Information
For this assay, SNP(s) [rs78184001] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HLA-DOB
Gene Name
major histocompatibility complex, class II, DO beta
There are no transcripts associated with this gene.

Gene
TAP2
Gene Name
transporter 2, ATP binding cassette subfamily B member
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000544.3 Intron NP_000535.3
NM_001290043.1 Intron NP_001276972.1
NM_018833.2 Intron NP_061313.2

View Full Product Details