Product Details

SNP ID
rs17764770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:162267800 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATAACTTCTTTCAAGTCTGTTGG[C/T]TATATTAACTTGACTTTAGTTGTTC
Phenotype
MIM: 606951
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFIH1 PubMed Links

Gene Details

Gene
IFIH1
Gene Name
interferon induced with helicase C domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022168.3 Intron NP_071451.2

View Full Product Details