Product Details

SNP ID
rs17376841
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:160152088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACAGTGGCTCCCCATGACCAGAG[C/T]CCAAGACGAAGACCTAAAAAAGGGC
Phenotype
MIM: 182340 MIM: 607321
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP1A2 PubMed Links

Gene Details

Gene
ATP1A2
Gene Name
ATPase Na+/K+ transporting subunit alpha 2
There are no transcripts associated with this gene.

Gene
ATP1A4
Gene Name
ATPase Na+/K+ transporting subunit alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001734.1 527 Intron NP_001001734.1
NM_144699.3 527 Silent Mutation AGC,AGT S16S NP_653300.2
XM_011509582.1 527 Intron XP_011507884.1

View Full Product Details