Product Details
- SNP ID
-
rs17136239
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:656343 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTTCCAAGATTTTTAAAATAACTT[C/T]GAATGGACACCACCTCCGTCTCTAT
- Phenotype
-
MIM: 615329
MIM: 609713
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EXOC2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs192257165] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EXOC2
- Gene Name
- exocyst complex component 2
- Gene
- HUS1B
- Gene Name
- HUS1 checkpoint clamp component B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_148959.3 |
622 |
Missense Mutation |
CAA,CGA |
Q201R |
NP_683762.2 |
View Full Product Details