Product Details

SNP ID
rs17856468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37698174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGTTCATGGCTACTATTAAAAG[C/G]TTTCCCACACTCTTCACACATATAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF607 PubMed Links

Gene Details

Gene
ZNF607
Gene Name
zinc finger protein 607
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172677.1 2421 Missense Mutation CCT,GCT P652A NP_001166148.1
NM_032689.4 2421 Missense Mutation CCT,GCT P653A NP_116078.4
XM_006723435.3 2421 Missense Mutation CCT,GCT P653A XP_006723498.1
XM_006723436.2 2421 Missense Mutation CCT,GCT P652A XP_006723499.1
XM_006723437.2 2421 Missense Mutation CCT,GCT P620A XP_006723500.1
XM_006723440.1 2421 Missense Mutation CCT,GCT P562A XP_006723503.1
XM_011527414.2 2421 Missense Mutation CCT,GCT P562A XP_011525716.1
Gene
ZNF781
Gene Name
zinc finger protein 781
There are no transcripts associated with this gene.

View Full Product Details