Product Details

SNP ID
rs17652610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:73157723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCATTACTTTTGTACAGGCTCTC[A/G]TTAATTCCACATAGATGGTCTAATG
Phenotype
MIM: 151690
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANXA1 PubMed Links

Gene Details

Gene
ANXA1
Gene Name
annexin A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000700.2 Intron NP_000691.1
XM_011518609.1 Intron XP_011516911.1
XM_017014657.1 Intron XP_016870146.1

View Full Product Details