Product Details
- SNP ID
-
rs17359286
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:102772987 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACCTGCTTGTACCTCATTTCCTCT[G/T]ATGGCCCAGAACTCATTTCCTATTG
- Phenotype
-
MIM: 185260
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MMP10
PubMed Links
Gene Details
- Gene
- MMP10
- Gene Name
- matrix metallopeptidase 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002425.2 |
1123 |
Silent Mutation |
ATA,ATC |
I362I |
NP_002416.1 |
View Full Product Details